Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Semaphorin 3A Rabbit anti-Human, Rat, Polyclonal, MilliporeSigma™

Catalog No. AB9604MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB9604MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB9604MI Supplier MilliporeSigma Supplier No. AB9604
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Semaphorin 3A Antibody, central region detects level of Semaphorin 3A and has been published and validated for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Semaphorin 3A
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Liquid in PBS containing 50% glycerol, 1mg/mL BSA and 0.05% sodium azide.
Gene Symbols SEMA3A; SEMAD; SEMAIII; SEMA1-PEN; semaphorin-like; MGC133243; SEMA1; Hsema-I; Hsema-III; SemD; coll-1; SEMAL
Host Species Rabbit
Immunogen Synthetic peptide from the central region of human semaphorin 3A.
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_006080.2
Target Species Human, Rat
Content And Storage −20°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.