Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Semaphorin 3F Rabbit anti-Human, Rat, Polyclonal, MilliporeSigma™

Catalog No. AB5471PMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB5471PMI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB5471PMI Supplier MilliporeSigma Supplier No. AB5471P
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Semaphorin 3F Antibody is an antibody against Semaphorin 3F for use in IC and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Semaphorin 3F
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Affinity Purified immunoglobulin. Liquid in 0.02M Phosphate buffer with 0.25M NaCl and 0.1% sodium azide.
Gene Symbols SEMA3F; SEMAK; SEMA-IV; SEMA4; Sema4; sema4
Host Species Rabbit
Immunogen Synthetic peptide from human Semaphorin 3F (Catalog number AG363). (TPWTQTQAVRFRFSRATDGALRPM).
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_004186.2
Target Species Human, Rat
Content And Storage 2-8°C in undiluted aliquotes for 12 months. Avoid repeated freeze/thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.