Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Semaphorin 4C, APC, Clone: 663203, R&D Systems™

Catalog No. FAB8497A025
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB8497A025 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB8497A025 Supplier R&D Systems Supplier No. FAB8497A025
This item is not returnable. View return policy

Specifications

Antigen Semaphorin 4C
Applications Flow Cytometry
Classification Monoclonal
Clone 663203
Conjugate APC
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. Q64151
Gene Alias FLJ20369, KIAA1739, MGC126382, MGC126383, M-Sema F, M-SEMA-F, sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, (semaphorin) 4C, SEMA4C, SEMACL1, SEMAF, SEMAI, semaphorin-4C
Gene Symbols SEMA4C
Host Species Rat
Immunogen Mouse myelona cell line NS0-derived recombinant mouse Semaphorin 4C, Glu22-Gly664, Accession # Q64151
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54910
Test Specificity Detects HEK293 human embryonic cell line transfected with mouse Semaphorin 4C by flow cytometry. Does not detect untransfected or irrelevant transfected HEK293 cells.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.