Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Semaphorin 4D/CD100 PE-conjugated Antibody, R&D Systems™
GREENER_CHOICESDP

Catalog No. FAB74701P Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB74701P 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB74701P Supplier R&D Systems Supplier No. FAB74701P
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 2 publications

Semaphorin 4D/CD100 Monoclonal specifically detects Semaphorin 4D/CD100 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Semaphorin 4D/CD100
Applications Flow Cytometry
Classification Monoclonal
Clone 758726
Conjugate R-PE
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. Q92854
Gene Alias A8, BB18, CD100, CD100 antigen, CD100M-sema-G, coll-4, FLJ33485, FLJ34282, FLJ39737, FLJ46484, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, 4D, MGC169138, MGC169141, M-sema-G, sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, (semaphorin) 4D, SEMA4D, SEMAJ, semaphorin-4D
Gene Symbols SEMA4D
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Semaphorin 4D/CD100 Met22-Arg734 Accession # Q92854
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10507
Test Specificity Detects human Semaphorin 4D/CD100 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human Semaphorin 4B, 4C, 4G, or recombinant mouse Semaphorin 4D is observed. In Western blots, no cross-reactivity with rhSemaphorin 4A, 6A, or 6C is observed.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.