Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

semenogelin I, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89016902 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-902 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-902 Supplier Abnova Corporation Supplier No. H00006406B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human SEMG1 protein.

The protein encoded by this gene is the predominant protein in semen. The encoded secreted protein is involved in the formation of a gel matrix that encases ejaculated spermatozoa. The prostate-specific antigen (PSA) protease processes this protein into smaller peptides, with each possibly having a separate function. The proteolysis process breaks down the gel matrix and allows the spermatozoa to move more freely. [provided by RefSeq

Sequence: MKPNIIFVLSLLLILEKQAAVMGQKGGSKGRLPSEFSQFPHGQKGQHYSGQKGKQQTESKGSFSIQYTYHVDANDHDQSRKSQQYDLNALHKTTKSQRHLGGSQQLLHNKQEGRDHDKSKGHFHRVVIHHKGGKAHRGTQNPSQDQGNSPSGKGISSQYSNTEERLWVHGLSKEQTSVSGAQKGRKQGGSQSSYVLQTEELVANKQQRETKNSHQNKGHYQNVVEVREEHSSKVQTSLCPAHQDKLQHGSKDIFSTQDELLVYNKNQHQTKNLNQDQQHGRKANKISYQSSSTEERRLHYGENGVQKDVSQRSIYSQTEKLVAGKSQIQAPNPKQEPWHGENAKGESGQSTNREQDLLSHEQKGRHQHGSHGGLDIVIIEQEDDSDRHLAQHLNNDQNPLFT

Specifications

Antigen semenogelin I
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human SEMG1 protein.
Formulation PBS with no preservative; pH 7.4
Gene SEMG1
Gene Accession No. BC007096.1
Gene Alias MGC14719/SEMG/SGI
Gene Symbols SEMG1
Host Species Mouse
Immunogen SEMG1 (AAH07096.1, 1 a.a. ∼ 402 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6406
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.