Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SENP6 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA511337
Description
SENP6 (SUSP1) is a protease that deconjugates SUMO1, SUMO2 and SUMO3 from targeted proteins. This protein does not seem to be involved in the processing of full-length SUMO proteins to their mature conjugatable forms. SENP6 deconjugates SUMO1 from RXRA, leading to transcriptional activation. It may act preferentially on substrates containing 3 or more SUMO2 or SUMO3 moieties.
Specifications
SENP6 | |
Polyclonal | |
Unconjugated | |
Q9GZR1 | |
SENP6 | |
KLH conjugated synthetic peptide between 1081-1112 amino acids from the C-terminal region of human SENP6 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
2 mg/mL | |
SENP6 | |
Sentrin-specific protease 6; Sentrin/SUMO-specific protease SENP6; SUMO-1-specific protease 1; SENP6; KIAA0797, SSP1, SUSP1 | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
26054 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction