Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

septin 11, Mouse, Clone: 1E5, Abnova™

Catalog No. 89001310 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-310 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-310 Supplier Abnova Corporation Supplier No. H00055752M02
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant SEPT11.

SEPT11 belongs to the conserved septin family of filament-forming cytoskeletal GTPases that are involved in a variety of cellular functions including cytokinesis and vesicle trafficking (Hanai et al., 2004 [PubMed 15196925]; Nagata et al., 2004 [PubMed 15485874]).[supplied by OMIM

Sequence: LRNLSLSGHVGFDSLPDQLVNKSTSQGFCFNILCVGETGIGKSTLMDTLFNTKFESDPATHNEPGVRLKARSYELQESNVRLKLTIVDTVGFGDQINKDDSYKPIVEYID

Specifications

Antigen Septin-11
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1E5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant SEPT11.
Formulation PBS with no preservative; pH 7.4
Gene SEPT11
Gene Accession No. BC008083
Gene Symbols SEPT11
Host Species Mouse
Immunogen SEPT11 (AAH08083, 71 a.a. ∼ 180 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Cycle
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 55752
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2b λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.