Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Serotonin 2A (5-HT2A) Receptor (22-41), Polyclonal

Catalog No. PC176100UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
PC176100UL 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PC176100UL Supplier MilliporeSigma™ Supplier No. PC176100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects Serotonin 2A (5-HT2A) Receptor (22-41) Clone: in Mouse, Rat, Sheep samples, and it is validated for Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immuno Blotting

At least 14 mammalian serotonin receptor subtypes have been identified and classified into several families on the basis of common structural, pharmacological, and functional criteria. Serotonin (5-hydroxytryptamine [5-HT]) mediates its effects in a number of physiological processes including anxiety, depression, sexual activity, and sleep through interactions with different receptor subtypes. These receptors differ in their tissue and cellular localization, affinity for serotonin, and second messenger pathways. The majority of these receptors stimulate a GTP-binding protein upon agonist stimulation and couple to adenylate cyclase or phospholipase C. In contrast, the serotonin 5-HT3 receptor acts as a cation-selective channel. The serotonin receptors have generated considerable pharmacological interest as targets for the identification of selective drugs that interact with a specific receptor subtype.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Serotonin 2A (5-HT2A) Receptor (22-41)
Applications Immunoblot, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Formulation In 150mM NaCl, 20mM sodium phosphate, 1% BSA, pH 7.5.
Host Species Rabbit
Immunogen a synthetic peptide corresponding to amino acids 22-41 of rat 5-HT2A receptor
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽53kDa serotonin 2A (5-HT2A) receptor.
Target Species Mouse, Rat, Sheep
Content And Storage −20°C
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.