Learn More
Serotonin Transporter Rabbit anti-Human, Rat, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Serotonin Transporter |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Affinity purified immunoglobulin. Liquid in 100mM Tris, pH 7.5 with 0.2% BSA and 0.05% sodium azide. |
| Gene Symbols | SLC6A4; 5-HTT; 5HTT; HTT; OCD1; hSERT; SERT |
| Host Species | Rabbit |
| Immunogen | A 15 amino acid sequence between transmembrane domain 7 and 8 of the rat serotonin transporter. This region is presumably from the extracellular domain. |
| Purification Method | Affinity Purified |
| Quantity | 50 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.