Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serotonin Transporter Mouse anti-Rat, Clone: 17-7A4, MilliporeSigma™

Catalog No. MAB1564MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1564MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1564MI Supplier MilliporeSigma Supplier No. MAB1564
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Serotonin Transporter using this Anti-Serotonin Transporter Antibody, clone 17-7A4 validated for use in immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Serotonin Transporter
Applications Immunohistochemistry
Classification Monoclonal
Clone 17-7A4
Conjugate Unconjugated
Formulation Protein A Purified mouse immunoglobulin in 20mM sodium phosphate, 250mM NaCl, pH. 7.6, with 0.1% sodium azide as a preservative.
Gene Symbols 5HTT, SC6A4, SLC6A4, HTT, SERT
Host Species Mouse
Immunogen Rat serotonin transporter, N-terminus/GST fusion protein (aa 1-85).
Purification Method Protein A purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Rat
Content And Storage 2-8°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.