Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serotonin Transporter Mouse anti-Human, Primate, MilliporeSigma™

Catalog No. MAB5618MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5618MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB5618MI Supplier MilliporeSigma Supplier No. MAB5618
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Serotonin Transporter Antibody detects level of Serotonin Transporter and has been published and validated for use in immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Serotonin Transporter
Applications Immunohistochemistry
Classification Monoclonal
Conjugate Unconjugated
Formulation Ascites
Gene Accession No. NM_001045.3
Gene Symbols SLC6A4, 5HTT, HTT, OCD1, hSERT, 5-HTT, SERT
Host Species Mouse
Immunogen Synthetic peptide from human serotonin transporter.
Purification Method Ascites
Quantity 100 μL
Primary or Secondary Primary
Target Species Human, Primate
Content And Storage −20°C in undiluted aliquots for 6 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.