Learn More
serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 1, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human SERPINA1 protein.
Supplier: Abnova Corporation H00005265D01P
Description
The protein encoded by this gene is secreted and is a serine protease inhibitor whose targets include elastase, plasmin, thrombin, trypsin, chymotrypsin, and plasminogen activator. Defects in this gene can cause emphysema or liver disease. Several transcript variants encoding the same protein have been found for this gene. [provided by RefSeq
Sequence: MPSSVSWGILLLAGLCCLVPVSLAEDPQGDAAQKTDTSHHDQDHPTFNKITPNLAEFAFSLYRQLAHQSNSTNIFFSPVSIATAFAMLSLGTKADTHDEILEGLNFNLTEIPEAQIHEGFQELLRTLNQPDSQLQLTTGNGLFLSEGLKLVDKFLEDVKKLYHSEAFTVNFGDTEEAKKQINDYVEKGTQGKIVDLVKELDRDTVFALVNYIFFKGKWERPFEVKDTEEEDFHVDQATTVKVPMMKRLGMFNIQHCKKLSSWVLLMKYLGNATAIFFLPDEGKLQHLENELTHDIITKFLENEDRRSASLHLPKLSITGTYDLKSVLGQLGITKVFSNGADLSGVTEEAPLKLSKAVHKAVLTIDEKGTEAAGAMFLEAIPMSIPPEVKFNKPFVFLMIEQNTKSPLFMGKVVNPTQKSpecifications
serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against a full-length human SERPINA1 protein. | |
SERPINA1 | |
A1A/A1AT/AAT/MGC23330/MGC9222/PI/PI1/PRO2275 | |
Rabbit | |
Affinity Purified | |
RUO | |
5265 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC015642.2 | |
SERPINA1 | |
SERPINA1 (AAH15642.1, 1 a.a. ∼ 418 a.a) full-length human protein. | |
100 μg | |
Primary | |
Human | |
Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.