Learn More
SERPINA3, Mouse, Clone: 1C10, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant SERPINA3.
Supplier: Abnova Corporation H00000012M02
Description
The protein encoded by this gene is a plasma protease inhibitor and member of the serine protease inhibitor class. Polymorphisms in this protein appear to be tissue specific and influence protease targeting. Variations in this protein's sequence have been implicated in Alzheimer's disease, and deficiency of this protein has been associated with liver disease. Mutations have been identified in patients with Parkinson disease and chronic obstructive pulmonary disease. [provided by RefSeq]
Sequence: PNSPLDEENLTQENQDRGTHVDLGLASANVDFAFSLYKQLVLKAPDKNVIFSPLSISTALAFLSLGAHNTTLTEILKGLKFNLTETSEAEIHQSFQHLLRTLNQSSDELQLSMGNAMFVKEQLSLLDRFTEDAKRLYGSEAFATDFQDSAAAKKLINDYVKNGTRGKITDLIKDLDSQTMMVLVNYIFFKAKWEMPFDPQDTHQSRFYLSKKKWVMVPMMSLHHLTIPYFRDEELSCTVVELKYTGNASALFILPDQDKMEEVEAMLLPETLKRWRDSLEFREIGELYLPKFSISRDYNLNDILLQLGIEEAFTSKADLSGITGARNLAVSQVVHKAVLDVFEEGTEASAATAVKITLLSALVETRTIVRFNRPFLMIIVPTDTQNIFFMSKVTNPKQASpecifications
SERPINA3 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC003559 | |
SERPINA3 | |
SERPINA3 (AAH03559, 25 a.a. ∼ 423 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed) | |
1C10 | |
Mouse monoclonal antibody raised against a full-length recombinant SERPINA3. | |
SERPINA3 | |
AACT/ACT/GIG24/GIG25/MGC88254 | |
Mouse | |
Affinity chromatography | |
RUO | |
12 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.