Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SERPINA6, Mouse, Clone: 1D9, Abnova™

Catalog No. 89016459 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-459 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-459 Supplier Abnova Corporation Supplier No. H00000866M02
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant SERPINA6.

This gene encodes an alpha-globulin protein with corticosteroid-binding properties. This is the major transport protein for glucorticoids and progestins in the blood of most vertebrates. The gene localizes to a chromosomal region containing several closely related serine protease inhibitors which may have evolved by duplication events. [provided by RefSeq

Sequence: MDPNAAYVNMSNHHRGLASANVDFAFSLYKHLVALSPKKNIFISPVSISMALAMLSLGTCGHTRAQLLQGLGFNLTERSETEIHQGFQHLHQLFAKSDTSLEMTMGNALFLDGSLELLESFSADIKHYYESEVLAMNFQDWATASRQINSYVKNKTQGKIVDLFSGLDSPAILVLVNYIFFKGTWTQPFDLASTREENFYVDETTVVKVPMMLQSSTISYLHDAELPCQLVQMNYVGNGTVFFILPDKGKMNTVIAALSRDTINRWSAGLTSSQVDLYIPKVTISGVYDLGDVLEEMGIADLFTNQANFSRITQDAQLKSSKVVHKAVLQLNEEGVDTAGSTGVTLNLTSKPIILRFNQPFIIMIFDHFTWSSLFLARVMNPV

Specifications

Antigen SERPINA6
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1D9
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant SERPINA6.
Formulation PBS with no preservative; pH 7.4
Gene SERPINA6
Gene Accession No. BC058021
Gene Alias CBG
Gene Symbols SERPINA6
Host Species Mouse
Immunogen SERPINA6 (AAH58021, 23 a.a. ∼ 405 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 866
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.