Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SERPINC1 (M02A), Mouse anti-Human, Clone: 2B12, Abnova™

Catalog No. 89016231 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-231 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-231 Supplier Abnova Corporation Supplier No. H00000462M02A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant SERPINC1.

The protein encoded by this gene is a plasma protease inhibitor and a member of the serpin superfamily. This protein inhibits thrombin as well as other activated serine proteases of the coagulation system, and it regulates the blood coagulation cascade. The protein includes two functional domains: the heparin binding-domain at the N-terminus of the mature protein, and the reactive site domain at the C-terminus. The inhibitory activity is enhanced by the presence of heparin. More than 120 mutations have been identified for this gene, many of which are known to cause antithrombin-III deficiency. [provided by RefSeq

Sequence: MYSNVIGTVTSGKRKVYLLSLLLIGFWDCVTCHGSPVDICTAEPRDIPMNPMCIYRSPEKKATEDEGSEQKIPEATNRRVWELSKANSRFATTFYQHLADSKNDNDNIFLSPLSISTAFAMTKLGACNDTLQQLMEVFKFDTISEKTSDQIHFFFAKLNCRLYRKANKSSKLVSANRLFGDKSLTFNDLYVSDAFHKAFLEVNEEGSEAAASTAVVIAGRSLNPNRVTFKANRPFLVFIREVPLNTIIFMGRVANPCVK

Specifications

Antigen SERPINC1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2B12
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant SERPINC1.
Formulation ascites with no preservative
Gene SERPINC1
Gene Accession No. BC022309
Gene Alias AT3/ATIII/MGC22579
Gene Symbols SERPINC1
Host Species Mouse
Immunogen SERPINC1 (AAH22309.1, 1 a.a. ∼ 259 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 462
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.