Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

serpin peptidase inhibitor, clade H (heat shock protein 47), member 1, (collagen binding protein 1), Mouse, Clone: 1D2-1A6, Abnova™

Catalog No. 89014068 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-014-068 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-014-068 Supplier Abnova Corporation Supplier No. H00000871M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant SERPINH1.

This gene encodes a member of the serpin superfamily of serine proteinase inhibitors. Its expression is induced by heat shock. The protein localizes to the endoplasmic reticulum lumen and binds collagen; thus it is thought to be a molecular chaperone involved in the maturation of collagen molecules. Autoantibodies to this protein have been found in patients with rheumatoid arthritis. [provided by RefSeq

Sequence: MRSLLLLSAFCLLEAALAAEVKKPAAAAAPGTAEKLSPKAATLAERSAGLAFSLYQAMAKDQAVENILVSPVVVASSLGLVSLGGKATTASQAKAVLSAEQLRDEEVHAGLGELLRSLSNSTARNVTWKLGSRLYGPSSVSFADDFVRSSKQHYNCEHSKINFRDKRSALQSINEWAAQTTDGKLPEVTKDVERTDGALLVNAMFFKPHWDEKFHHKMVDNRGFMVTRSYTVGVMMMHRTGLYNYYDDEKEKLQIVEMPLAHKLSSLIILMPHHVEPLERLEKLLTKEQLKIWMGKMQKKAVAISLPKGVVEVTHDLQKHLAGLGLTEAIDKNKADLSRMSGKKDLYLASVFHATAFELDTDGNPFDQDIYGREELRSPKLFYADHPFIFLVRDTQSGSLLFIGRLVRPKGDKMRDEL

Specifications

Antigen serpin peptidase inhibitor, clade H (heat shock protein 47), member 1, (collagen binding protein 1)
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1D2-1A6
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant SERPINH1.
Formulation PBS with no preservative; pH 7.4
Gene SERPINH1
Gene Accession No. BC014623
Gene Alias AsTP3/CBP1/CBP2/HSP47/PIG14/PPROM/RA-A47/SERPINH2/gp46
Gene Symbols SERPINH1
Host Species Mouse
Immunogen SERPINH1 (AAH14623.1, 1 a.a. ∼ 418 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Stem Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 871
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.