Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SESN1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA526881
Description
Sestrin-1 is involved in the reduction of peroxiredoxins. This protein may also be regulator of cellular growth.
Specifications
SESN1 | |
Polyclonal | |
SESN1 | |
PA26, SEST1, p53 activated gene 26, p53 regulated PA26 nuclear protein, sestrin-1 | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
140742, 27244 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
Unconjugated | |
Q9Y6P5, P58006 | |
SESN1 | |
KLH conjugated synthetic peptide between 14-43 amino acids from the N-terminal region of human Sestrin-1 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction