Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SETD7 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA511283
Description
Histone methyltransferases (HMTases) selectively methylate evolutionarily conserved arginine or lysine residues, primarily in the N-terminal tails of histones H3 and H4. Signal transduction pathways affecting the N-terminal tails of histones lead to a number of post-translational modifications including acetylation, phosphorylation, poly(ADP-ribosylation), ubiquitination and methylation. These modifications play critical roles in regulating chromatin structure and gene expression. Set7/9 is a histone specific HMTase that methylates histone H3 lysine 4. Set7/9 transfers methyl groups to lysine 4 of histone H3 in complex with S-adenosyl-L-methionine. In yeast, H4-K20 methylation does not have any apparent role in the regulation of gene expression or heterochromatin function; rather it appears to play a role in DNA damage response. Loss of Set9 activity or mutation of H4-K20 markedly impairs yeast cell survival after genotoxic challenge and compromises the ability of cells to maintain checkpoint mediated cell cycle arrest. Genetic experiments link Set9 to Crb2, a homolog of the mammalian checkpoint protein 53BP1, and the enzyme is required for Crb2 localization to sites of DNA damage.
Specifications
SETD7 | |
Polyclonal | |
SETD7 | |
Histone H3-K4 methyltransferase; H3-K4-HMTase; SET domain-containing protein 7; Set9; SET7/9;; KMT7 | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
73251, 80854 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
Unconjugated | |
Q8WTS6, Q8VHL1 | |
SETD7 | |
KLH conjugated synthetic peptide between 289-317 amino acids from the C-terminal region of human SET7 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction