Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SF-1 Rabbit anti-Mouse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma 07618
Description
Anti-SF-1 Antibody is a high quality Rabbit Polyclonal Antibody for the detection of SF-1 and has been validated in western blotting.
Specifications
SF-1 | |
Polyclonal | |
Protein A Purified immunoglobulin in 30% glycerol, 0.07M Tris-glycine, pH 7.4, 0.105M NaCl, 0.035% sodium azide as a preservative. | |
Rabbit | |
Protein A purfied | |
Primary | |
Mouse | |
Purified |
Western Blot | |
Unconjugated | |
NR5A1; FTZ1; FTZF1; AD4BP; SF-1; STF-1; OTTHUMP00000042845; SF1; ELP; OTTHUMP00000042847; OTTHUMP00000042846 | |
DNA binding domain of SF-1 from mouse, produced in E. coli as a GST fusion protein and partially purified via GST-agarose column chromatography | |
200 μg | |
NM_004959 | |
−20°C in undiluted aliquotes for 24 months. Avoid repeated freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction