Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-SF3B2, Polyclonal, Abnova™

Catalog No. 89300483 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-300-483 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-300-483 Supplier Abnova Corporation Supplier No. H00010992B01P
Add to Cart
Add to Cart

Mouse Polyclonal Antibody

Specifications

Antigen SF3B2
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human SF3B2 protein.
Formulation PBS with no preservative; pH 7.4
Gene SF3B2
Gene Accession No. BC007610.1
Gene Alias SAP145/SF3B145/SF3b1/SF3b150
Gene Symbols SF3B2
Host Species Mouse
Immunogen SF3B2 (AAH07610.1, 1 a.a. ∼ 636 a.a) full-length human protein.
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10992
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.