Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SFTPB, Rabbit, Monoclonal Antibody, Abnova™

Catalog No. 89210156 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg x up to 3 Abs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-210-156 1 mg x up to 3 Abs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-210-156 Supplier Abnova Corporation Supplier No. H00006439K

Includes

Up to three rabbit IgG clones of 1mg each will be delivered to customer.

Rabbit monoclonal antibody raised against a human SFTPB peptide using ARM Technology.

The SFTPB gene encodes the pulmonary-associated surfactant B protein (SPB), an amphipathic surfactant protein essential for lung function and homeostasis after birth. Pulmonary surfactant is a lipid-rich material that prevents lung collapse by lowering surface tension at the air-liquid interface in the alveoli of lung. SPB enhances the rate of spreading and increases the stability of surfactant monolayers in vitro. Surfactant is composed of phospholipids and other surfactant-associated proteins (Clark et al., 1995 [PubMed 7644495]). See also SFTPA1 (MIM 178630), SFTPC (MIM 178620), and SFTPD (MIM 178635).[supplied by OMIM

Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)
Expression: Overexpression vector and transfection into 293H cell line

Order Info

  1. Customer may provide cell or tissue lysate for antibody screening.
  2. Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.

Specifications

Antigen SFTPB
Applications ELISA, Western Blot
Classification Monoclonal
Concentration 1mg
Conjugate Unconjugated
Description surfactant protein B
Formulation In 1X PBS, pH 7.4
Gene SFTPB
Gene Accession No. SFTPB
Gene Alias PSP-B, SFTB3, SFTP3, SMDP1, SP-B
Gene Symbols SFTPB
Host Species Rabbit
Immunogen A synthetic peptide of human SFTPB
Quantity 1 mg x up to 3 Abs
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6439
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.