Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

surfactant protein B, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89006224 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-006-224 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-006-224 Supplier Abnova Corporation Supplier No. H00006439B01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human SFTPB protein.

The SFTPB gene encodes the pulmonary-associated surfactant B protein (SPB), an amphipathic surfactant protein essential for lung function and homeostasis after birth. Pulmonary surfactant is a lipid-rich material that prevents lung collapse by lowering surface tension at the air-liquid interface in the alveoli of lung. SPB enhances the rate of spreading and increases the stability of surfactant monolayers in vitro. Surfactant is composed of phospholipids and other surfactant-associated proteins (Clark et al., 1995 [PubMed 7644495]). See also SFTPA1 (MIM 178630), SFTPC (MIM 178620), and SFTPD (MIM 178635).[supplied by OMIM

Sequence: MAESHLLQWLLLLLPTLCGPGTAAWTTSSLACAQGPEFWCQSLEQALQCRALGHCLQEVWGHVGADDLCQECEDIVHILNKMAKEAIFQDTMRKFLEQECNVLPLKLLMPQCNQVLDDYFPLVIDYFQNQTDSNGICMHLGLCKSRQPEPEQEPGMSDPLPKPLRDPLPDPLLDKLVLPVLPGALQARPGPHTQDLSEQQFPIPLPYCWLCRALIKRIQAMIPKGALAVAVAQVCRVVPLVAGGICQCLAERYSVILLDTLLGRMLPQLVCRLVLRCSMDDSAGPRSPTGEWLPRDSECHLCMSVTTQAGNSSEQAIPQAMLQACVGSWLDREKCKQFVEQHTPQLLTLVPRGWDAHTTCQALGVCGTMSSPLQCIHSPDL

Specifications

Antigen surfactant protein B
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human SFTPB protein.
Formulation No additive
Gene SFTPB
Gene Accession No. NM_000542.2
Gene Alias PSP-B/SFTB3/SFTP3/SMDP1/SP-B
Gene Symbols SFTPB
Host Species Mouse
Immunogen SFTPB (NP_000533.2, 1 a.a. ∼ 381 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 6439
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.