Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

sideroflexin 2, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89008497 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-008-497 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-008-497 Supplier Abnova Corporation Supplier No. H00118980B01

Mouse polyclonal antibody raised against a full-length human SFXN2 protein.

Sequence: MEADLSGFNIDAPRWDQRTFLGRVKHFLNITDPRTVFVSERELDWAKVMVEKSRMGVVPPGTQVEQLLYAKKLYDSAFHPDTGEKMNVIGRMSFQLPGGMIITGFMLQFYRTMPAVIFWQWVNQSFNALVNYTNRNAASPTSVRQMALSYFTATTTAVATAVGMNMLTKKAPPLVGRWVPFAAVAAANCVNIPMMRQQELIKGICVKDRNENEIGHSRRAAAIGITQVVISRITMSAPGMILLPVIMERLEKLHFMQKVKVLHAPLQVMLSGCFLIFMVPVACGLFPQKCELPVSYLEPKLQDTIKAKYGELEPYVYFNKGL

Specifications

Antigen sideroflexin 2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description sideroflexin 2
Formulation No additive
Gene SFXN2
Gene Accession No. BC022091
Gene Symbols SFXN2
Host Species Mouse
Immunogen SFXN2 (AAH22091, 1 a.a. ∽323 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 118980
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.