Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SG2NA Mouse anti-Human, Rat, Mouse, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 051115
Description
Anti-SG2NA Antibody is an antibody against SG2NA for use in western blot, IC and immunoprecipitation.
Specifications
SG2NA | |
Monoclonal | |
Mouse monoclonal IgG1 ascites containing 40% glycerol and 0.02% sodium azide. | |
STRN3, Striatin-3, GS2NA, SG2NA | |
Synthetic peptide from SG2NA. | |
Primary | |
−20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles. | |
IgG1 |
Immunocytochemistry, Immunoprecipitation, Western Blot | |
Unconjugated | |
NM_001083893.1 | |
Mouse | |
100 μL | |
Human, Mouse, Rat | |
Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction