Learn More
SGCA, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human SGCA peptide using ARM Technology.
Supplier: Abnova Corporation H00006442K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
This gene encodes a component of the dystrophin-glycoprotein complex (DGC), which is critical to the stability of muscle fiber membranes and to the linking of the actin cytoskeleton to the extracellular matrix. Its expression is thought to be restricted to striated muscle. Mutations in this gene result in type 2D autosomal recessive limb-girdle muscular dystrophy. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
SGCA | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
SGCA | |
SGCA | |
A synthetic peptide of human SGCA | |
RUO | |
6442 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
sarcoglycan, alpha (50kDa dystrophin-associated glycoprotein) | |
SGCA | |
50-DAG, A2, ADL, DAG2, DMDA2, LGMD2D, SCARMD1, adhalin | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.