Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SH2D2A Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA525250
Description
This protein encodes an adaptor protein thought to function in T-cell signal transduction. A related protein in mouse is responsible for the activation of lymphocyte-specific protein-tyrosine kinase and functions in downstream signaling. Alternative splicing results in multiple transcript variants.
Specifications
SH2D2A | |
Polyclonal | |
SH2D2A | |
F2771, SCAP, TSAD, VRAP, SH2 domain protein 2A, SH2 domain-containing protein 2A, T cell specific adapter protein TSAd, T lymphocyte specific adaptor protein, VEGF receptor-associated protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9047 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
Unconjugated | |
Q9NP31 | |
SH2D2A | |
KLH conjugated synthetic peptide between 86-112 amino acids from the central region of human SH2D2A | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction