Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SH3-domain binding protein 4, Mouse, Clone: 2B6, Abnova™

Catalog No. 89001234 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-234 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-234 Supplier Abnova Corporation Supplier No. H00023677M11
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant SH3BP4.

This gene encodes a protein with 3 Asn-Pro-Phe (NPF) motifs, an SH3 domain, a PXXP motif, a bipartite nuclear targeting signal, and a tyrosine phosphorylation site. This protein is involved in cargo-specific control of clathrin-mediated endocytosis, specifically controlling the internalization of a specific protein receptor. [provided by RefSeq

Sequence: MAAQRIRAANSNGLPRCKSEGTLIDLSEGFSETSFNDIKVPSPSALLVDNPTPFGNAKEVIAIKDYCPTNFTTLKFSKGDHLYVLDTSGGEWWYAHNTTEMGYIPSSYVQ

Specifications

Antigen SH3-domain binding protein 4
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2B6
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant SH3BP4.
Formulation PBS with no preservative; pH 7.4
Gene SH3BP4
Gene Accession No. BC057396
Gene Alias BOG25/TTP
Gene Symbols SH3BP4
Host Species Mouse
Immunogen SH3BP4 (AAH57396, 1 a.a. ∼ 110 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Cycle
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 23677
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.