Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SH3KBP1, Rabbit, Monoclonal Antibody, Abnova™

Catalog No. 89202284 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg x up to 3 Abs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-202-284 1 mg x up to 3 Abs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-202-284 Supplier Abnova Corporation Supplier No. H00030011K

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Includes

Up to three rabbit IgG clones of 1mg each will be delivered to customer.

Rabbit monoclonal antibody raised against a human SH3KBP1 peptide using ARM Technology.

CBL (MIM 165360) constitutively interacts with SH3 domain-containing proteins and, upon tyrosine phosphorylation, with SH2 domain-containing proteins. The SH3KBP1 gene encodes an 85kDa CBL-interacting protein that enhances tumor necrosis factor (MIM 191160)-mediated apoptotic cell death (Narita et al., 2001 [PubMed 11474197]).[supplied by OMIM

Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)
Expression: Overexpression vector and transfection into 293H cell line

Order Info

  1. Customer may provide cell or tissue lysate for antibody screening.
  2. Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.

Specifications

Antigen SH3KBP1
Applications ELISA, Western Blot
Classification Monoclonal
Concentration 1mg
Conjugate Unconjugated
Description SH3-domain kinase binding protein 1
Formulation In 1X PBS, pH 7.4
Gene SH3KBP1
Gene Accession No. SH3KBP1
Gene Alias CIN85, GIG10, MIG18
Gene Symbols SH3KBP1
Host Species Rabbit
Immunogen A synthetic peptide of human SH3KBP1
Quantity 1 mg x up to 3 Abs
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30011
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.