Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHANK1a Rabbit anti-Rat, Polyclonal, MilliporeSigma™

Catalog No. AB5852MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB5852MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB5852MI Supplier MilliporeSigma Supplier No. AB5852
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-SHANK1a Antibody, N-terminus is an antibody against SHANK1a for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SHANK1a
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid in PBS containing 0.02% sodium azide.
Gene Symbols SHANK1; SSTRIP; SPANK-1; Shank1; synamon
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the N-terminus of rat Shank1a.
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_016148.2
Target Species Rat
Content And Storage −20°C in undiluted aliquotes for 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.