Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-Sheep Red Blood Cells, Polyclonal, IgG Fraction, MP Biomedicals
Supplier: MP Biomedicals, Inc 0855806
Description
Host Species: Rabbit
Isotype: IgG
Species Reactivity: Sheep

Specifications
Red blood cells | |
Unconjugated | |
2 mL | |
Lyophilized |
Polyclonal | |
Rabbit | |
Primary | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction