Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHP-1/SHPTP-1 Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. 07419MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-419-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-419-MI Supplier MilliporeSigma Supplier No. 07419
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-SHP-1/SHPTP-1 Antibody is validated for use in immunoprecipitation, western blotting for the detection of SHP-1/SHPTP-1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SHP-1/SHPTP-1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation 0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide
Gene Symbols PTPN6; HCP; SH-PTP1; PTP-1C; SHP1; SHP-1L; PTP1C; HCPH; SHP-1; HPTP1C
Host Species Rabbit
Immunogen Full-length glutathione transferase SHP-1 fusion protein, purified from bacteria
Purification Method Protein A purfied
Quantity 200 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_002831
Target Species Human
Content And Storage 2-8°C for 12 months
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.