Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHP2 Mouse, Unlabeled, Clone: 79, BD

Catalog No. BDB610621 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610621 50 μg
BDB610622 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610621 Supplier BD Biosciences Supplier No. 610621
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SHP2 (also known as PTP1D, Syp, SHPTP2 and PTP2C) is a member of the cytosolic class of protein-tyrosine phosphatases (PTPs). SHP2 has been reported to contain two SH2 domains where both SH2 domains are N-terminal to the PTP catalytic domain. The expression of SHP2 has been reported to be highest in brain, heart, and kidney. The PTPs are thought to function with other protein-tyrosine kinases to maintain intracellular protein phosphotyrosine homeostasis and cell cycle progression. The presence ofSH2 domains in SHP2 (PTP1D) has prompted speculation that binding to specific phosphorylated tyrosine residues is key to its function. SHP2 (PTP1D) is tyrosine- phosphorylated and activated in response to stimulation with EGF or PDGF.

TRUSTED_SUSTAINABILITY

Specifications

Antigen SHP2
Applications Western Blot
Classification Monoclonal
Clone 79
Concentration 250μg/mL
Conjugate Unconjugated
Description PTP1D, Syp, SHPTP2 and PTP2C
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Chicken, Frog, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.