Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHP2 (pY542) Mouse, PE, Clone: L99-921, BD

Catalog No. BDB560389 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560389 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB560389 Supplier BD Biosciences Supplier No. 560389
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SHP2 is a member of the cytosolic class of protein-tyrosine phosphatases (PTPs). SHP2 reportedly contains two SH2 domains, both of which are N-terminal to the PTP catalytic domain. SH2 PTPs are believed to work in conjunction with protein-tyrosine kinases to maintain intracellular protein phosphotyrosine homeostasis and cell cycle progression. The expression of SHP2 has been reported to be highest in brain, heart, and kidney. SHP2 is phosphorylated at two C-terminal tyrosine residues: Y542 and Y580. Phosphorylation of Y542 creates an interaction in the N-terminal SH2 domain and appears to relieve basal inhibition of the PTP, while phosphorylation at Y580 creates an interaction at the C-terminal that stimulates PTP activity. The L99-921 monoclonal antibody recognizes the phosphorylated Y542 of SHP2 (PTP2C isoform 2). The homologous phosphorylation site in the PTP2Ci splice variant (isoform 1) is Y546. The specificity of this antibody was validated by confirming, using western blot analysis, that RNA-mediated interference (RNAi) of the specific protein was able to down-regulate the expression of SHP2 (pY542).

Host Species: Mouse
Clone: L99-921
Isotype: IgG1 κ
Species Reactivity: Human
Immunogen: Phosphorylated Human SHP2 Peptide

Intracellular Staining

TRUSTED_SUSTAINABILITY

Specifications

Antigen SHP2 (pY542)
Applications Flow Cytometry
Classification Monoclonal
Clone L99-921
Conjugate PE
Description PTN11, PTP1D, PTP2C, PTPN11, SH-PTP2, SH-PTP3, SHP-2, SHPTP2
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Phosphorylated Human SHP2 Peptide
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Rat
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.