Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHPS-1 Rat anti-Mouse, Clone: P84, MilliporeSigma™

Catalog No. MABS164MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS164MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS164MI Supplier MilliporeSigma Supplier No. MABS164

Rat Monoclonal Antibody

Anti-SHPS-1 Antibody, clone P84 detects level of SHPS-1 and has been published and validated for use in western blot, immunocytochemistry, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SHPS-1
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone P84
Conjugate Unconjugated
Formulation Purified rat monoclonal IgG1k in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_001171117
Gene Symbols Sirpa, Bit, Myd1, Ptpns1, Shps1, Sirp
Host Species Rat
Immunogen Linear peptide corresponding to mouse SHPS-1.
Purification Method Protein G Purified
Quantity 100 μL
Primary or Secondary Primary
Target Species Mouse
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.