Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

single immunoglobulin and toll-interleukin 1 receptor (TIR) domain, Mouse, Clone: 3H8-2G3, Abnova™

Catalog No. 89001549 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-549 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-549 Supplier Abnova Corporation Supplier No. H00059307M01A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant SIGIRR.

Sequence: MPGVCDRAPDFLSPSEDQVLRPALGSSVALNCTAWVVSGPHCSLPSVQWLKDGLPLGIGGHYSLHEYSWVKANLSEVLVSSVLGVNVTSTEVYGAFTCSIQNISFSSFTLQRAGPTSHVAAVLASLLVLLALLLAALLYVKCRLNVLLWYQDAYGEVEINDGKLYDAYVSYSDCPEDRKFVNFILKPQLERRRGYKLFLDDRDLLPRAEPSADLLVNLSRCRRLIVVLSDAFLSRAWCSHSFREGLCRLLELTRRPIFITFEGQRRDPAHPALRLLRQHRHLVTLLLWRPGSVTPSSDFWKEVQLALPRKVRYRPVEGDPQTQLQDDKDPMLILRGRVPEGRALDSEVDPDPEGDLGVRGPVFGEPSAPPHTSGVSLGESRSSEVDVSDLGSRNYSARTDFYCLVSKDDM

Specifications

Antigen single immunoglobulin and toll-interleukin 1 receptor (TIR) domain
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3H8-2G3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant SIGIRR.
Formulation ascites with no preservative
Gene SIGIRR
Gene Accession No. BC003591
Gene Alias MGC110992/TIR8
Gene Symbols SIGIRR
Host Species Mouse
Immunogen SIGIRR (AAH03591, 1 a.a. ∼ 410 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 59307
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.