Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-SIGIRR Clone: 3H8-2G3,

Catalog No. ST1681200UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ST1681200UL 200 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ST1681200UL Supplier MilliporeSigma™ Supplier No. ST1681200UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects SIGIRR Clone: 3H8-2G3 in Human samples, and it is validated for Immuno Blotting

The SIGIRR protein acts as a negative regulator of the Toll-like and IL-1R receptor signaling pathways. It attenuates the recruitment of receptor-proximal signaling components to the TLR4 receptor, probably through an TIR-TIR domain interaction with TLR4. Its extracellular domain is known to interfere with the heterodimerization of Il1R1 and IL1RAP.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIGIRR
Applications Immunoblot
Classification Monoclonal
Clone 3H8-2G3
Conjugate Unconjugated
Formulation Undiluted ascites
Host Species Mouse
Immunogen Full-length, recombinant, human SIGIRR (aa 1-411), expressed as a GST fusion protein
Purification Method Unpurified
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽32kDa SIGIRR protein in HepG2 cells.
Target Species Human
Content And Storage −20°C
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.