Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Siglec-1 (CD169) Clone: 5F1.1,

Catalog No. MABT328MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABT328MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABT328MI Supplier MilliporeSigma™ Supplier No. MABT328
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects Siglec-1 (CD169) Clone: 5F1.1 in Human, Mouse samples, and it is validated for Immunohistochemistry (Paraffin), Western Blotting

Sialoadhesin (UniProt Q9BZZ2; also known as CD169, Sialic acid-binding Ig-like lectin 1, Sialic acid-binding immunoglobulin-like lectin 1, Siglec-1) is encoded by the SIGLEC1 (also known as SN) gene (Gene ID 6614) in human. Sialoadhesin/CD169/Siglec-1 is the founding member of the sialic acid-binding immunoglobulin lectins (Siglec) expressed on a variety of leukocytes and stromal cells. Sialoadhesin is initially produced with a signal sequence (a.a. 1-19), the removal of which yields the mature protein with a large extracellular portion (a.a. 20-1641), followed by a transmembrane segment (a.a. 1642-1662), and a short cytoplasmic tail (a.a. 1663-1709) with no known signal transduction or endocytosis motifs. The extracellular portion contains an N-terminal Ig-like V-type domain (a.a. 20-136), followed by sixteen Ig-like C2-type domains. Sialoadhesin is highly expressed on the subcapsular sinus (SCS) and medullary macrophages in lymph node (LN) and on marginal metallophilic macrophages in the marginal zone (MZ) of spleen. Although sialic acids are found on the surface of all cells and most secreted proteins, Only heavily sialylated, multimeric structures are captured by Sialoadhesin on macrophages via its N-terminal Ig-like V-type domain due to a low (millimolar) affinity toward sialylic acid. Sialoadhesin-mediated sialic-acid recognition allows macrophages to engage sialylated pathogens and stimulate inflammatory responses. Sialoadhesin is also reported to capture of B cell-derived exosomes via their surface-expressed α2, 3-linked sialic acids. In the lymph node, exosomes are not retained in the subcapsular sinus of Siglec1-knockout mice and penetrate deeper into the paracortex, causing an enhanced response to antigen-pulsed exosomes among Siglec1-/- mice.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Siglec-1 (CD169)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 5F1.1
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2aκ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q9BZZ2
Gene Symbols SIGLEC1, SN
Host Species Mouse
Immunogen GST-tagged recombinant human Siglec-1 (CD169) N-terminal fragment containing the Ig-like V-type domain.
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Structure
Primary or Secondary Primary
Gene ID (Entrez) NP_075556
Test Specificity Expected to react with all three spliced isoforms of human Siglec-1 (CD169)/Sialoadhesin reported by UniProt (Q9BZZ2).
Target Species Human, Mouse
Content And Storage 2°C to 8°C for one year from date of shipment
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.