Learn More
Human Siglec-3/CD33 APC-conjugated Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Supplier:  R&D Systems FAB1137A100
 View more versions of this product
Catalog No. FAB1137A100
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
Siglec-3/CD33 Monoclonal specifically detects Siglec-3/CD33 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
Siglec-3/CD33
Monoclonal
APC
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
CD33
Recombinant human Siglec-3/CD33 Extracellular domain
100 Tests
Primary
Detects human Siglec-3/CD33 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Siglec-2, -5, -7, or -9 is observed.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
IgG1
Flow Cytometry
6C5/2
Flow Cytometry 10 uL/10^6 cells
CD33, CD33 antigen, CD33 antigen (gp67), CD33 molecule, FLJ00391, gp67, myeloid cell surface antigen CD33, p67, sialic acid binding Ig-like lectin 3, Sialic acid-binding Ig-like lectin 3, Siglec3, SIGLEC-3, SIGLEC3gp67
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
945
Human
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.