Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Siglec-8 Alexa Fluor 700-conjugated Antibody, R&D Systems™
GREENER_CHOICESDP

Catalog No. FAB7975N Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB7975N 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB7975N Supplier R&D Systems Supplier No. FAB7975N

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Siglec-8 Monoclonal specifically detects Siglec-8 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Siglec-8
Applications Flow Cytometry
Classification Monoclonal
Clone 837535
Conjugate Alexa Fluor 700
Dilution Flow Cytometry 5 uL/10^6 cells
Gene Accession No. Q9NYZ4
Gene Alias CD329 antigen, CDw329, MGC59785, SAF2, SAF2SAF-2, sialic acid binding Ig-like lectin 8, Sialoadhesin family member 2, Siglec8, Siglec-8, SIGLEC8L, SIGLEC8Lsialic acid-binding Ig-like lectin 8
Gene Symbols SIGLEC8
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Siglec-8 Met17-Ala363 Accession # Q9NYZ4
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Primary or Secondary Primary
Gene ID (Entrez) 27181
Test Specificity Detects human Siglec-8 in ELISAs. In direct ELISAs, no cross-reactivity with recombinant human Siglec-7, -9, or -14 is observed.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.