Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Siglec-8 Mouse anti-Human, Clone: 7C9, MilliporeSigma™

Catalog No. 041473MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-147-3MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-147-3MI Supplier MilliporeSigma Supplier No. 041473

Mouse Monoclonal Antibody

Detect Siglec-8 using this Anti-Siglec-8 Antibody, clone 7C9 validated for use in flow cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Siglec-8
Applications Flow Cytometry
Classification Monoclonal
Clone 7C9
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgGκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_055257
Gene Symbols SIGLEC8, CDw329, SAF2, SAF-2, SIGLEC8L, SIGLEC-8, Siglec-8
Host Species Mouse
Immunogen Extracellular region of human recombinant Siglec-8 cleaved with Factor Xa.
Purification Method Protein G Purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.