Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Siglec-9 PE-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB1139P025
This item is not returnable.
View return policy
Description
Siglec-9 Monoclonal specifically detects Siglec-9 in Human samples. It is validated for Flow Cytometry.Specifications
| Siglec-9 | |
| Monoclonal | |
| R-PE | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CD329, CDw329, FOAP-9, OBBP-Like, Protein FOAP-9, sialic acid binding Ig-like lectin 9, sialic acid-binding Ig-like lectin 9, siglec-9, Siglec9 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 27180 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 191240 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Q9Y336 | |
| SIGLEC9 | |
| Mouse myeloma cell line NS0-derived recombinant human Siglec-9 Gln18-Gly348 (predicted) Accession # Q9Y336 | |
| 25 Tests | |
| Primary | |
| Detects human Siglec-9 in ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human Siglec-2, -3, -5, -7 or -10 is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction