Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Siglec-G Mouse anti-Mouse, Alexa Fluor 647, Clone: SH1, BD

Catalog No. BDB563336 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB563336 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB563336 Supplier BD Biosciences Supplier No. 563336
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The SH1 monoclonal antibody specifically binds to mouse Siglec-G (sialic acid binding immunoglobulin-like lectin G), also known as Siglec10. Siglec-G is a Type I transmembrane glycoprotein that belongs to the Immunoglobulin superfamily. Siglec-G functions as an adhesion molecule that mediates sialic-acid dependent binding to cells. Siglec-G is expressed on cells of the B cell lineage. Siglec-G is most highly expressed by pre-B cells and B1a cells within the B cell lineage and is not detectable on T cells. Siglec-G can inhibit B cell receptor-mediated calcium signaling when it is overexpressed. Mice that lack Siglec-G have significantly increased numbers of B1a cells that begins early in development and is B cell intrinsic. Siglec-G-deficient mice have higher titers of natural IgM antibodies than their normal counterparts. Mouse Siglec-G is the ortholog of human Siglec-10 (also known as, CD330).

TRUSTED_SUSTAINABILITY

Specifications

Antigen Siglec-G
Applications Flow Cytometry
Classification Monoclonal
Clone SH1
Concentration 0.2mg/mL
Conjugate Alexa Fluor 647
Description Siglecg; mSiglec-G; Siglec10; Siglec 10; CD330
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Mouse Extracellular Siglec-G Recombinant Protein
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.