Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sigma IR Rat anti-Human, Mouse, Clone: ICR-SIG1R-A, MilliporeSigma™
Rat Monoclonal Antibody
Supplier: MilliporeSigma MABN390
Description
Detect the Sigma IR protein using this Anti-Sigma IR, clone ICR-SIG1R-A validated for use in western blot, immunoprecipitation, immunocytochemistry, Flow and immunohistochemistry.
Specifications
Sigma IR | |
Monoclonal | |
Unconjugated | |
NP_005857 | |
Rat | |
Protein G purified | |
Primary | |
2-8°C for one year | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
ICR-SIG1R-A | |
Purified rat monoclonal IgG1 in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
SIGMAR1, OPRS1, SRBP | |
Linear peptide corresponding to human Sigma IR. | |
100 μg | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction