Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIII p15 Mouse, Unlabeled, Clone: 56, BD

Catalog No. BDB610761 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610761 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB610761 Supplier BD Biosciences Supplier No. 610761

Mouse Monoclonal Antibody

SIII p15 is one of three polypeptides that comprise the approximate 140kDa RNA polymerase II transcription factor SIII. The other two polypeptides have the molecular weights of 110kDa and 18kDa. All three polypeptides are required for transcription factor SIII function. Although the polypeptides were identified and purified from rat liver nuclear extracts, tissue expression of this transcription factor appears to be widespread. RNA polymerase II recognizes the SIII transcription factor and in turn initiates transcription from these correctly initiated transcripts. A yeast transcription factor designated YES may be the yeast homolog to SIII.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen SIII p15
Applications Western Blot
Classification Monoclonal
Clone 56
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human SIII p15 aa. 1-112
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Frog, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.