Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIK1 Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. ABE799MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE799MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE799MI Supplier MilliporeSigma Supplier No. ABE799
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-SIK1 Antibody detects level of SIK1 and has been published and validated for use in SIK1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIK1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.
Gene Symbols SIK1; SIK; SNF1LK
Host Species Rabbit
Immunogen KLH-conjugated synthetic peptide corresponding to the center region of human SIK1.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.