Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

single-minded homolog 2 (Drosophila), Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89004310 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-004-310 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-004-310 Supplier Abnova Corporation Supplier No. H00006493A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant SIM2.

SIM1 and SIM2 genes are Drosophila single-minded (sim) gene homologs. The Drosophila sim gene encodes a transcription factor that is a master regulator of fruit fly neurogenesis. SIM2 maps within the so-called Down syndrome chromosomal region. Based on the mapping position, its potential function as transcriptional repressor and similarity to Drosophila sim, it is proposed that SIM2 may contribute to some specific Down syndrome phenotypes [provided by RefSeq

Sequence: ESSDLLYTPSYSLPFSYHYGHFPLDSHVFSSKKPMLPAKFGQPQGSPCEVARFFLSTLPASGECQWHYANPLVPSSSSPAKNPPEPPANTARHSLVPSYE

Specifications

Antigen single-minded homolog 2 (Drosophila)
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant SIM2.
Formulation 50% glycerol
Gene SIM2
Gene Accession No. NM_005069
Gene Alias MGC119447/SIM/bHLHe15
Gene Symbols SIM2
Host Species Mouse
Immunogen SIM2 (NP_005060, 426 a.a. ∼ 525 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Research Discipline Neurobiology
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 6493
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.