Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sin1 Mouse anti-Human, Mouse, Rat, Clone: 1C7.2, MilliporeSigma™

Catalog No. 051044MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-104-4MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-104-4MI Supplier MilliporeSigma Supplier No. 051044
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Sin1 using this Anti-Sin1 Antibody, clone 1C7.2 validated for use in western blot, immunoprecipitation, immunohistochemistry (paraffin) and immunofluoresence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Sin1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 1C7.2
Conjugate Unconjugated
Formulation Purified mouse monoclonal in 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% NaN3.
Gene Accession No. NP_001006618
Gene Symbols JC310, MGC2745, MIP1, SIN1
Host Species Mouse
Immunogen Full-length Human Sin1 GST-fusion protein.
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.