Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Sin3A, Polyclonal

Catalog No. 06913IMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
06-913-IMI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-913-IMI Supplier MilliporeSigma™ Supplier No. 06913I
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects Sin3A Clone: in Human, Mouse samples, and it is validated for Western Blotting

Paired amphipathic helix protein Sin3a (UniProt Q60520; also known as Histone deacetylase complex subunit Sin3a, mSin3a, Transcriptional co-repressor Sin3a) is encoded by the Sin3A (also known as mSin3A, AW553200, Kiaa4126, mKIAA4126) gene (Gene ID 8819) in murine species. The transcription regulator Sin3a is a scaffold protein that interacts with various proteins to form specialized complexes that mediate different biological processes, including chromatin modification, embryonic development, DNA repair, mitochondrial metabolism, and gene transcription. SIN3a harbors four pairedα-helices (PAH), a histone interaction domain, and a highly conserved region. The histone interaction domain mediates HDACs binding, whereas the highly conserved region interacts with proteins necessary for repression, such as the nuclear receptor hormone co-repressor Alien. The four PAHs represent the main interacting domains of SIN3a and mediate the interactions with a wide variety of DNA-binding transcription factors. Members of the Mad family of repressor proteins, for example, specifically interact with the PAH2 through their Sin3 interacting domain (SID). Although widely studied for its involvement in transcriptional repression, Sin3a is shown to be essential for aryl hydrocarbon receptor-mediated induction of gene transcription.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Sin3A
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q60520
Gene Symbols Sin3a, Kiaa4126
Host Species Rabbit
Immunogen KLH-conjugated peptide (KRRLDDQESPVYAAQQRR-C) corresponding to amino acids 2-19 of mouse Sin3A.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) NP_035508
Target Species Human, Mouse
Content And Storage 2°C to 8°C for one year
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.