Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIP1 Mouse anti-Human, Clone: 6E5, MilliporeSigma™

Catalog No. MABD407MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABD407MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABD407MI Supplier MilliporeSigma Supplier No. MABD407
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-SIP1 Antibody, clone 6E5 is an antibody against SIP1 for use in western blotting, immunocytochemistry and immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIP1
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 6E5
Concentration 1mg/mL
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2aκ supernatant in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_001165124
Gene Symbols ZEB2, KIAA0569, SIP1, ZFHX1B, ZFX1B, HRIHFB2411
Host Species Mouse
Immunogen His-tagged recombinant protein corresponding to the N-terminus of Human SIP1.
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.