Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SIPA1L2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA520848
Description
A suggested positive control is rat brain tissue lysate. PA5-20848 can be used with blocking peptide PEP-0962.
Signal-induced proliferation associated-like protein 2 (SIPA1L2) is a member of the SIPA1 family of RapGAPs. Little is known of the role of the SIPA1L2 protein, but recent studies of SIPA indicate that its deregulation can cause myeloproliferative stem cell disorders in mice and increased metastases in human cancers. Other studies suggest SIPA1L1 may play important roles in embryo development and control of cell proliferation. Based on the amount of homology between SIPA family members, it is likely that SIPA1L2 plays a role in embryo development and cell proliferation, possibly including oncogenesis.
Specifications
SIPA1L2 | |
Polyclonal | |
Unconjugated | |
SIPA1L2 | |
BC058408; Kiaa0545; KIAA1389; mKIAA1389; serine-rich synapse associated protein 2; serine-rich synapse-associated protein; Sersap2; signal induced proliferation associated 1 like 2; signal-induced proliferation-associated 1 like 2; signal-induced proliferation-associated 1-like protein 2; Sipa1l2; SIPA1-like protein 2; SPA-1-like 2; SPAL2; Spar2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
244668, 361442, 57568 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q5JCS6, Q80TE4, Q9P2F8 | |
SIPA1L2 | |
A 19 amino acid peptide from near the amino terminus of human SIPA1L2. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction